Assay Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146241.2 | NP_666353.1 | 12 - 13 | 2260 | 113 | |
| GenBank mRNA | AK029833.1 | - | 1590 | 113 | ||
| AK034093.1 | - | 12 - 13 | 1639 | 113 | ||
| BC032288.1 | - | 12 - 13 | 2265 | 5368580 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trhde
Genomic Map