Assay Details
| Gene Symbol: Trhde | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146241.2 | NP_666353.1 | 8 - 9 | 1791 | 142 | |
| GenBank mRNA | AK029833.1 | - | 1121 | 142 | ||
| AK034093.1 | - | 8 - 9 | 1170 | 142 | ||
| BC032288.1 | - | 8 - 9 | 1796 | 5368580 | 142 | |
More Information
Set Membership: |
|

Trhde
Genomic Map