Assay Details
| Gene Symbol: Pitpnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024927.2 | NP_001020098.1 | 3 - 4 | 444 | 69 | |
| GenBank mRNA | AK039594.1 | - | 3 - 4 | 333 | 69 | |
| AK147444.1 | - | 3 - 4 | 401 | 69 | ||
| BC112411.1 | - | 2 - 3 | 819 | 40061037 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm3
Genomic Map