Assay Details
| Gene Symbol: Pitpnm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024927.2 | NP_001020098.1 | 6 - 7 | 800 | 60 | |
| NM_001081641.1 | NP_001075110.1 | 5 - 6 | 752 | 60 | ||
| GenBank mRNA | AK039594.1 | - | 5 - 6 | 993 | 60 | |
| AK147444.1 | - | 6 - 7 | 757 | 60 | ||
| AK147630.1 | - | 5 - 6 | 585 | 60 | ||
| BC112411.1 | - | 5 - 6 | 1175 | 40061037 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pitpnm3
Genomic Map