Assay Details
| Gene Symbol: Mlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026810.2 | NP_081086.2 | 4 - 5 | 458 | 79 | |
| GenBank mRNA | AF250844.1 | - | 4 - 5 | 374 | 79 | |
| AK038265.1 | - | 4 - 5 | 451 | 79 | ||
| AK145646.1 | - | 4 - 5 | 404 | 79 | ||
| AK163861.1 | - | 4 - 5 | 392 | 79 | ||
| AK168849.1 | - | 4 - 5 | 397 | 79 | ||
| BC021815.1 | - | 4 - 5 | 412 | 4020489 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlh1
Genomic Map