Assay Details
| Gene Symbol: Mlh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026810.2 | NP_081086.2 | 8 - 9 | 760 | 72 | |
| GenBank mRNA | AF250844.1 | - | 8 - 9 | 676 | 72 | |
| AK038265.1 | - | 8 - 9 | 753 | 72 | ||
| AK145646.1 | - | 8 - 9 | 707 | 72 | ||
| AK168849.1 | - | 8 - 9 | 699 | 72 | ||
| BC021815.1 | - | 8 - 9 | 714 | 4020489 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mlh1
Genomic Map