Assay Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083331.2 | NP_001076800.1 | 8 - 9 | 1313 | 126 | |
| NM_001276406.1 | NP_001263335.1 | 8 - 9 | 1313 | 126 | ||
| NM_172394.3 | NP_765982.2 | 8 - 9 | 1313 | 126 | ||
| GenBank mRNA | AJ532593.1 | - | 8 - 9 | 1307 | 126 | |
| AK002643.1 | - | 8 - 9 | 1299 | 126 | ||
| AK028563.1 | - | 8 - 9 | 1303 | 126 | ||
| AK039007.1 | - | 8 - 9 | 1303 | 126 | ||
| AK050893.1 | - | 8 - 9 | 1302 | 126 | ||
| AK085776.1 | - | 8 - 9 | 1312 | 126 | ||
| AK160619.1 | - | 8 - 9 | 1314 | 126 | ||
| AK166214.1 | - | 8 - 9 | 1303 | 126 | ||
| BC032929.1 | - | 8 - 9 | 1294 | 1379673 | 126 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup88
Genomic Map