Assay Details
| Gene Symbol: Nup88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083331.2 | NP_001076800.1 | 9 - 10 | 1406 | 144 | |
| NM_001276406.1 | NP_001263335.1 | 9 - 10 | 1406 | 144 | ||
| NM_172394.3 | NP_765982.2 | 9 - 10 | 1406 | 144 | ||
| GenBank mRNA | AJ532593.1 | - | 9 - 10 | 1400 | 144 | |
| AK002643.1 | - | 9 - 10 | 1392 | 144 | ||
| AK028563.1 | - | 9 - 10 | 1396 | 144 | ||
| AK050893.1 | - | 9 - 10 | 1395 | 144 | ||
| AK085776.1 | - | 9 - 10 | 1405 | 144 | ||
| AK160619.1 | - | 9 - 10 | 1407 | 144 | ||
| AK166214.1 | - | 9 - 10 | 1396 | 144 | ||
| BC032929.1 | - | 9 - 10 | 1387 | 1379673 | 144 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Nup88
Genomic Map