Assay Details
| Gene Symbol: Gm31718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050164.1 | - | 7 - 8 | 740 | 69 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interferon regulatory factor 2 binding protein 2 | Irf2bp2 |
E130305N23Rik, IRF-2BP2 |
-221.0kb | Sense | Chr. 8 - 126588296 - 126593436 on NCBI Build GRCm38 |
| Downstream | RIKEN cDNA A630001O12 gene | A630001O12Rik | 29.8kb | Anti-sense | Chr. 8 - 126848899 - 126852254 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm31718
Genomic Map