Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 17 - 18 | 1788 | 93 | |
| GenBank mRNA | AF017176.1 | - | 14 - 15 | 1436 | 93 | |
| AK076037.1 | - | 17 - 18 | 1749 | 93 | ||
| AK146334.1 | - | 12 - 13 | 1261 | 93 | ||
| BC026559.1 | - | 17 - 18 | 1767 | 5037878 | 93 | |
| U41497.1 | - | 8 - 9 | 93 | |||
| Z71189.1 | - | 17 - 18 | 1709 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
very long-chain fatty acid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
oxidation-reduction process
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
long-chain-acyl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
very-long-chain-acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding
temperature homeostasis
lipid metabolic process
fatty acid metabolic process
fatty acid beta-oxidation
metabolic process
fatty acid beta-oxidation using acyl-CoA dehydrogenase
very long-chain fatty acid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
oxidation-reduction process
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
long-chain-acyl-CoA dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-CH group of donors
very-long-chain-acyl-CoA dehydrogenase activity
flavin adenine dinucleotide binding
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991906 | BC092396.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991906 | Z71189.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991906 | AF017176.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991906 | AK076037.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1991906 | NM_017366.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 854885 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 934627 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 752694 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145484 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991906 | BC092396.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991906 | Z71189.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991906 | AF017176.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991906 | AK076037.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1991906 | NM_017366.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518340 | BC092396.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518340 | Z71189.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518340 | AK146334.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518340 | BC026559.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P518340 | AF017176.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898511 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898514 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898530 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898533 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898546 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898539 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898538 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898505 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898536 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898507 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898516 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898548 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898545 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898550 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898517 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898541 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898527 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898534 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898508 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27898531 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P145484 | - | - |
| Whole Mouse Genome G4122A | A_51_P518340 | BC092396.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518340 | Z71189.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518340 | AK146334.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518340 | BC026559.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518340 | AF017176.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113286 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P145484 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09479681 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666710 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666707 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666708 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09479685 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09479695 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09479691 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666686 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09666689 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09479679 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09233284 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09233273 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P09233281 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898420 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898541 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898559 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898600 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898490 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898536 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898545 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898502 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898462 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898550 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898572 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898577 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898589 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898582 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898498 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898557 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898422 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898562 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898434 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898459 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898599 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898495 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898602 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898597 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898496 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898426 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898456 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898543 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898429 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27898552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898488 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666704 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666695 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666702 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898511 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666708 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898530 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898490 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09479693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898516 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666699 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898523 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P09666689 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898508 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P27898534 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10387788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387782 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387786 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387769 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387772 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387770 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387777 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387781 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387773 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387790 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387775 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387771 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387776 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387778 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387780 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387783 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387774 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10387779 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424184_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10387768 | - | - |
| Mouse Genome 430 2.0 Array | 1424184_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424184_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11370_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11370_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1100028394.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028394.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028397.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028397.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028399.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028399.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028392.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028392.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028395.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028395.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028400.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028400.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028393.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028393.mm.1 | TC1100003083.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1100028401.mm.1 | NM_017366.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1100028401.mm.1 | TC1100003083.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424184_at | - | - |
| Mouse Expression Set 430 | 1424184_PM_at | - | - |
| Murine Genome U74v2 Set | 161623_at | - | - |
| Murine Genome U74v2 Set | 161623_PM_at | - | - |
| Murine Genome U74v2 Set | 93334_PM_at | - | - |
| Murine Genome U74v2 Set | 93334_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424184_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424184_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424184_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424184_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6040403 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6040403 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006040403 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2775586 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2775586 | - | - |
| mouse-6 v1.1 (Illumina) | scl40004.19.1_140-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6040403 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl40004.19.1_140-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6040403 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2956932 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2775586 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2956932 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl40004.19.1_140-S | - | - |

Acadvl
Genomic Map