Assay Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017366.3 | NP_059062.1 | 1 - 2 | 168 | 93 | |
| GenBank mRNA | AK076037.1 | - | 1 - 2 | 129 | 92 | |
| AU067294.1 | - | 1 - 2 | 168 | 93 | ||
| BC026559.1 | - | 1 - 2 | 147 | 5037878 | 93 | |
| Z71189.1 | - | 1 - 2 | 89 | 93 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Acadvl
Genomic Map