Assay Details
| Gene Symbol: Pitpnc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB077282.1 | - | 8 - 9 | 740 | 67 | |
| AK003844.1 | - | 7 - 8 | 710 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Pitpnc1
Genomic Map