Assay Details
| Gene Symbol: Slc26a11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178743.3 | NP_848858.2 | 15 - 16 | 1734 | 57 | |
| GenBank mRNA | AF345196.1 | - | 15 - 16 | 1734 | 57 | |
| AK052202.1 | - | 10 - 11 | 1821 | 57 | ||
| AK089182.1 | - | 15 - 16 | 1714 | 57 | ||
| BC038604.1 | - | 15 - 16 | 1727 | 57 | ||
| BC132493.1 | - | 10 - 11 | 1602 | 40130770 | 57 | |
| BC137897.1 | - | 10 - 11 | 1602 | 8860916 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc26a11
Genomic Map