Assay Details
| Gene Symbol: March10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039242.2 | NP_001034331.1 | 1 - 2 | 268 | 131 | |
| NM_172568.2 | NP_766156.2 | 1 - 2 | 268 | 131 | ||
| GenBank mRNA | AK132734.1 | - | 1 - 2 | 257 | 131 | |
| BC100474.1 | - | 1 - 2 | 261 | 30918723 | 131 | |
| BY099161.1 | - | 1 - 2 | 269 | 131 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK132734.1, BC100474.1, NM_001039242.2, NM_172568.2 transcripts.
Set Membership: |
|

March10
Genomic Map