Assay Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001167680.1 | NP_001161152.1 | 7 - 8 | 1106 | 96 | |
| NM_172572.3 | NP_766160.2 | 7 - 8 | 1320 | 96 | ||
| GenBank mRNA | AK028872.1 | - | 7 - 8 | 1321 | 96 | |
| AK042365.1 | - | 6 - 7 | 1773 | 96 | ||
| AK154639.1 | - | 7 - 8 | 1086 | 96 | ||
| AK171085.1 | - | 7 - 8 | 1102 | 96 | ||
| AY167043.1 | - | 7 - 8 | 1106 | 96 | ||
| BC031398.1 | - | 1 - 2 | 139 | 96 | ||
| BC052182.1 | - | 7 - 8 | 1054 | 6335022 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rhbdf2
Genomic Map