Assay Details
| Gene Symbol: BC100451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021440.2 | NP_067415.2 | 1 - 2 | 225 | 80 | |
| GenBank mRNA | BC100451.1 | - | 1 - 2 | 225 | 30918483 | 80 |
| Y09878.1 | - | 1 - 2 | 191 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC100451.1, NM_021440.2, Y09878.1 transcripts.
Set Membership: |
|

BC100451
Genomic Map