Assay Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033528.1 | NP_001028700.1 | 11 - 12 | 1567 | 60 | |
| GenBank mRNA | AK028572.1 | - | 11 - 12 | 1612 | 60 | |
| AK049205.1 | - | 11 - 12 | 1397 | 60 | ||
| AK129363.1 | - | 11 - 12 | 1365 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp36
Genomic Map