Assay Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033528.1 | NP_001028700.1 | 14 - 15 | 2476 | 90 | |
| GenBank mRNA | AK028572.1 | - | 14 - 15 | 2522 | 91 | |
| AK049205.1 | - | 14 - 15 | 2306 | 90 | ||
| AK129363.1 | - | 14 - 15 | 2274 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp36
Genomic Map