Assay Details
| Gene Symbol: Usp36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033528.1 | NP_001028700.1 | 8 - 9 | 1227 | 63 | |
| GenBank mRNA | AK028572.1 | - | 8 - 9 | 1272 | 63 | |
| AK049205.1 | - | 8 - 9 | 1057 | 63 | ||
| AK129363.1 | - | 8 - 9 | 1025 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Usp36
Genomic Map