Assay Details
| Gene Symbol: 4930506A18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK015712.1 | - | 2 - 3 | 173 | 158 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
sphingomyelin phosphodiesterase 3, neutral |
Smpd3 |
4631433G07Rik, AI427456, AW537966, fro, nSMase2 |
Anti-sense | Chr. 8 - 106252548 - 106337988 on NCBI Build GRCm38 | |
| Downstream | zinc finger protein 90 | Zfp90 |
6430515L01Rik, KRAB17, NK10, Zfp64, Zfp83, mKIAA1954 |
80.9kb | Sense | Chr. 8 - 106415284 - 106425889 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

4930506A18Rik
Genomic Map