Assay Details
| Gene Symbol: Gm14169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040372.1 | 3 - 4 | 494 | 141 | ||
| GenBank mRNA | AK032272.1 | - | 3 - 4 | 495 | 141 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | AAR2 splicing factor homolog (S. cerevisiae) | Aar2 | 0610011L14Rik | -40.2kb | Anti-sense | Chr. 2 - 156547558 - 156568972 on NCBI Build GRCm38 |
| Downstream |
discs, large homolog-associated protein 4 (Drosophila) |
Dlgap4 |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
1.1kb | Anti-sense | Chr. 2 - 156613633 - 156764363 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm14169
Genomic Map