Assay Details
| Gene Symbol: AU023762 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040760.1 | 2 - 3 | 172 | 89 | ||
| GenBank mRNA | AK049838.1 | - | 2 - 3 | 173 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 36367 |
Gm36367 | -190.3kb | Anti-sense | Chr. 9 - 113287712 - 113310985 on NCBI Build GRCm38 | |
| Downstream |
predicted gene, 36745 |
Gm36745 | 113.3kb | Sense | Chr. 9 - 113635860 - 113644603 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

AU023762
Genomic Map