Assay Details
| Gene Symbol: 2900079G21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033433.1 | 2 - 3 | 330 | 71 | ||
| GenBank mRNA | AK047318.1 | - | 2 - 3 | 330 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

2900079G21Rik
Genomic Map