Assay Details
| Gene Symbol: Gm26843 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK038973.1 | - | 2 - 3 | 760 | 98 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | carboxylesterase 5A | Ces5a |
1700081L16Rik, 1700122C07Rik, BB081581, Ces7, Gm503, cauxin |
-214.3kb | Sense | Chr. 8 - 93499063 - 93535881 on NCBI Build GRCm38 |
| Downstream |
guanine nucleotide binding protein, alpha O |
Gnao1 |
AW050213, Galphao, Gnao, alphaO |
1.7kb | Anti-sense | Chr. 8 - 93810561 - 93969388 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm26843
Genomic Map