Assay Details
| Gene Symbol: Gm12320 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039373.1 | - | 1 - 2 | 197 | 99 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | kinesin family member 1C | Kif1c |
B430105J22Rik, D11Bwg1349e, Ltxs1 |
-18.3kb | Anti-sense | Chr. 11 - 70700548 - 70731970 on NCBI Build GRCm38 |
| Downstream | zinc finger protein 3 | Zfp3 |
AI854635, B230309J24, Fnp-1, Zfp-3 |
9.3kb | Anti-sense | Chr. 11 - 70764447 - 70772928 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm12320
Genomic Map