Assay Details
| Gene Symbol: Tnrc6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198022.2 | NP_932139.2 | 13 - 14 | 4542 | 63 | |
| GenBank mRNA | AK036989.1 | - | 9 - 10 | 2021 | 63 | |
| AK122529.1 | - | 9 - 10 | 2493 | 63 | ||
| AK147471.1 | - | 14 - 15 | 4540 | 63 | ||
| AK147551.1 | - | 13 - 14 | 4258 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc6c
Genomic Map