Assay Details
| Gene Symbol: Tnrc6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198022.2 | NP_932139.2 | 11 - 12 | 4287 | 65 | |
| GenBank mRNA | AK036989.1 | - | 7 - 8 | 1766 | 65 | |
| AK122529.1 | - | 7 - 8 | 2238 | 65 | ||
| AK147471.1 | - | 12 - 13 | 4285 | 65 | ||
| AK147551.1 | - | 11 - 12 | 4003 | 65 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tnrc6c
Genomic Map