Assay Details
| Gene Symbol: Tha1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027919.4 | NP_082195.2 | 3 - 4 | 534 | 95 | |
| GenBank mRNA | AY219871.1 | - | 3 - 4 | 534 | 95 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tha1
Genomic Map