Assay Details
| Gene Symbol: Casp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009807.2 | NP_033937.2 | 1 - 2 | 208 | 138 | |
| GenBank mRNA | AK132826.1 | - | 1 - 2 | 40 | 138 | |
| AK163069.1 | - | 1 - 2 | 39 | 138 | ||
| BC008152.1 | - | 1 - 2 | 208 | 3583883 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC008152.1, NM_009807.2 transcripts.
Set Membership: |
|

Casp1
Genomic Map