Assay Details
| Gene Symbol: Sin3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009188.4 | NP_033214.2 | 13 - 14 | 2366 | 93 | |
| GenBank mRNA | AK087970.1 | - | 6 - 7 | 1463 | 93 | |
| AK159076.1 | - | 13 - 14 | 2349 | 93 | ||
| AK173005.1 | - | 12 - 13 | 4316 | 93 | ||
| BC129859.1 | - | 13 - 14 | 2338 | 93 | ||
| L38622.2 | - | 13 - 14 | 2338 | 93 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sin3b
Genomic Map