Assay Details
| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005746.1 | - | 4 - 5 | 305 | 98 | |
| AK046224.1 | - | 4 - 5 | 308 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp4r1l-ps| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK005746.1 | - | 4 - 5 | 305 | 98 | |
| AK046224.1 | - | 4 - 5 | 308 | 98 | ||
Set Membership: |
|