Assay Details
| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027957.1 | 12 - 13 | 2235 | 101 | ||
| GenBank mRNA | AK029171.1 | - | 13 - 14 | 4515 | 101 | |
| AK035093.1 | - | 4 - 5 | 1876 | 101 | ||
| AK041597.1 | - | 5 - 6 | 1473 | 101 | ||
| AK046224.1 | - | 11 - 12 | 2132 | 101 | ||
| AK047187.1 | - | 4 - 5 | 1326 | 101 | ||
| AK161459.1 | - | 11 - 12 | 2338 | 101 | ||
| AK164611.1 | - | 12 - 13 | 2237 | 101 | ||
| AK170078.1 | - | 11 - 12 | 1319 | 101 | ||
| BC064312.1 | - | 4 - 5 | 701 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp4r1l-ps
Genomic Map