Assay Details
| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_027957.1 | 13 - 14 | 2347 | 95 | ||
| GenBank mRNA | AK021059.1 | - | 2 - 3 | 141 | 95 | |
| AK029171.1 | - | 14 - 15 | 4627 | 95 | ||
| AK164611.1 | - | 13 - 14 | 2349 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp4r1l-ps
Genomic Map