Assay Details
| Gene Symbol: Irf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050834.1 | - | 147 | 80 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 17296 |
Gm17296 |
Gm179, Tarbp1 |
-91.9kb | Anti-sense | Chr. 8 - 126425314 - 126475113 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Irf2bp2
Genomic Map