Assay Details
| Gene Symbol: Irf2bp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK050834.1 | - | 211 | 97 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
predicted gene, 17296 |
Gm17296 |
Gm179, Tarbp1 |
-97.5kb | Anti-sense | Chr. 8 - 126425314 - 126475113 on NCBI Build GRCm38 |
| Downstream |
predicted gene, 31718 |
Gm31718 | 214.4kb | Anti-sense | Chr. 8 - 126804707 - 126908515 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Irf2bp2
Genomic Map