Assay Details
| Gene Symbol: 9330175E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015514.2 | 1 - 2 | 270 | 94 | ||
| GenBank mRNA | AK053303.1 | - | 1 - 2 | 272 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330175E14Rik| Gene Symbol: 9330175E14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015514.2 | 1 - 2 | 270 | 94 | ||
| GenBank mRNA | AK053303.1 | - | 1 - 2 | 272 | 94 | |
Set Membership: |
|