Assay Details
| Gene Symbol: Herpud1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022331.1 | NP_071726.1 | 2 - 3 | 324 | 75 | |
| GenBank mRNA | AB034991.1 | - | 2 - 3 | 324 | 75 | |
| AK159500.1 | - | 2 - 3 | 330 | 75 | ||
| AK165832.1 | - | 2 - 3 | 330 | 75 | ||
| AK172874.1 | - | 2 - 3 | 317 | 75 | ||
| AK186384.1 | - | 1 - 2 | 75 | |||
| BC013523.1 | - | 2 - 3 | 316 | 4236803 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Herpud1
Genomic Map