Assay Details
| Gene Symbol: Amfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011787.2 | NP_035917.2 | 10 - 11 | 1547 | 63 | |
| GenBank mRNA | AF124144.1 | - | 10 - 11 | 1755 | 63 | |
| AK038442.1 | - | 10 - 11 | 1515 | 63 | ||
| AK157327.1 | - | 10 - 11 | 1553 | 63 | ||
| AK170716.1 | - | 10 - 11 | 1533 | 63 | ||
| AK199083.1 | - | 1 - 2 | 59 | 63 | ||
| AK201825.1 | - | 1 - 2 | 99 | 63 | ||
| AK202389.1 | - | 1 - 2 | 99 | 63 | ||
| BC003256.1 | - | 2 - 3 | 185 | 63 | ||
| BC034538.1 | - | 10 - 11 | 1505 | 4209258 | 63 | |
| BC040338.1 | - | 10 - 11 | 1473 | 3710780 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amfr
Genomic Map