Assay Details
| Gene Symbol: Amfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011787.2 | NP_035917.2 | 12 - 13 | 1767 | 85 | |
| GenBank mRNA | AF124144.1 | - | 12 - 13 | 1975 | 85 | |
| AK038442.1 | - | 12 - 13 | 1735 | 85 | ||
| AK157327.1 | - | 12 - 13 | 1773 | 85 | ||
| AK170716.1 | - | 12 - 13 | 1753 | 85 | ||
| AK194945.1 | - | 1 - 2 | 85 | 85 | ||
| AK199083.1 | - | 3 - 4 | 279 | 85 | ||
| BC003256.1 | - | 4 - 5 | 405 | 85 | ||
| BC034538.1 | - | 12 - 13 | 1725 | 4209258 | 85 | |
| BC040338.1 | - | 12 - 13 | 1693 | 3710780 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|

Amfr
Genomic Map