Assay Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205311.1 | NP_001192240.1 | 15 - 16 | 1954 | 90 | |
| NM_019415.2 | NP_062288.2 | 15 - 16 | 1951 | 90 | ||
| GenBank mRNA | AK052691.1 | - | 15 - 16 | 1955 | 90 | |
| AK052755.1 | - | 15 - 16 | 1953 | 90 | ||
| AK085496.1 | - | 15 - 16 | 1960 | 90 | ||
| BC038612.1 | - | 15 - 16 | 1976 | 4237274 | 90 | |
| U61085.1 | - | 15 - 16 | 1933 | 90 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a3
Genomic Map