Assay Details
| Gene Symbol: Slc12a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205311.1 | NP_001192240.1 | 7 - 8 | 983 | 74 | |
| NM_019415.2 | NP_062288.2 | 7 - 8 | 980 | 74 | ||
| GenBank mRNA | AK052691.1 | - | 7 - 8 | 984 | 74 | |
| AK052755.1 | - | 7 - 8 | 982 | 74 | ||
| AK085496.1 | - | 7 - 8 | 987 | 74 | ||
| BC038612.1 | - | 7 - 8 | 1005 | 4237274 | 74 | |
| U61085.1 | - | 7 - 8 | 962 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc12a3
Genomic Map