Assay Details
| Gene Symbol: Gnao1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010308.3 | NP_034438.1 | 6 - 7 | 1221 | 83 | |
| GenBank mRNA | AK035907.1 | - | 6 - 7 | 1222 | 83 | |
| AK047197.1 | - | 6 - 7 | 1225 | 83 | ||
| BC051989.1 | - | 6 - 7 | 1048 | 5686124 | 83 | |
| M36777.1 | - | 6 - 7 | 744 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gnao1
Genomic Map