Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 14 - 15 | 1695 | 69 | |
| GenBank mRNA | AK134274.1 | - | 14 - 15 | 1696 | 69 | |
| AK138442.1 | - | 14 - 15 | 1719 | 69 | ||
| AK146469.1 | - | 14 - 15 | 1723 | 69 | ||
| BC011321.1 | - | 13 - 14 | 1572 | 4007058 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx27
Genomic Map