Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 19 - 20 | 2209 | 62 | |
| GenBank mRNA | AK134274.1 | - | 19 - 20 | 2210 | 62 | |
| AK138442.1 | - | 19 - 20 | 2233 | 62 | ||
| AK146469.1 | - | 19 - 20 | 2237 | 62 | ||
| AK209187.1 | - | 4 - 5 | 363 | 62 | ||
| BC011321.1 | - | 18 - 19 | 2086 | 4007058 | 62 | |
| BC026381.1 | - | 4 - 5 | 398 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21833121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21833119 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748411 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 821710 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 216979 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 484773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 512002 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 527640 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 546430 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 493379 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK044451.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK083517.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK085725.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK150227.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK158139.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AK190052.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | AY223875.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | BC013261.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | BC026611.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | BC033273.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | BC058620.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 530448 | NM_146217.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 616889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 175941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 14805 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 34544 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P161285 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164730 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833127 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833120 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833116 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833109 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833132 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833124 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833121 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833126 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833128 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833114 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833130 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833123 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833119 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833112 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833134 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833129 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21833122 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P161285 | - | - |
| Whole Mouse Genome G4122A | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P138772 | BC011321.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240671 | BC024981.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240671 | BC033398.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P240671 | BC024730.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103335 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P161285 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02243629 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02243627 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02243633 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21833121 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P21833119 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833147 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833130 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833138 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833136 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833156 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833151 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833086 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833120 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833139 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833122 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833129 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833121 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833149 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833109 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833123 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833107 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02243627 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833144 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833120 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833099 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833117 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833127 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833103 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833157 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02255535 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833091 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02195089 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01712381 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02015341 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833151 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833132 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02201879 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833098 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02201880 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833085 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833156 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02243630 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833104 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833114 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02195090 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833112 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10478826 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478844 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478845 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478843 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478828 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478842 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478841 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478833 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478846 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10478830 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1424161_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1425541_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478825 | - | - |
| Mouse Genome 430 2.0 Array | 1424161_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1424161_at | - | - |
| Mouse Genome 430 2.0 Array | 1425541_at | - | - |
| Mouse Genome 430 2.0 Array | 1425541_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228889_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 228889_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200021239.mm.1 | NM_153065.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021239.mm.1 | NM_153065.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021239.mm.1 | TC0200002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200021242.mm.1 | NM_153065.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021242.mm.1 | NM_153065.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021242.mm.1 | TC0200002579.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0200021241.mm.1 | NM_153065.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021241.mm.1 | NM_153065.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200021241.mm.1 | TC0200002579.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1424161_PM_at | - | - |
| Mouse Expression Set 430 | 1424161_at | - | - |
| Mouse Expression Set 430 | 1425541_PM_at | - | - |
| Mouse Expression Set 430 | 1425541_at | - | - |
| Murine Genome U74v2 Set | 95540_PM_r_at | - | - |
| Murine Genome U74v2 Set | 95540_r_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425541_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424161_at | - | - |
| Mouse Genome 430A 2.0 Array | 1424161_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1425541_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424161_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425541_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1425541_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1424161_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6980594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6980594 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 107040026 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006980594 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2634582 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2634582 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2507890 | - | - |
| mouse-6 v1.1 (Illumina) | scl00228889.2_16-S | - | - |
| mouse-6 v1.1 (Illumina) | scl19891.17.1_25-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6980594 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 107040026 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl00228889.2_16-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 107040026 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6980594 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2944784 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2634582 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2507890 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2944784 | - | - |

Ddx27
Genomic Map