Assay Details
| Gene Symbol: Ddx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153065.3 | NP_694705.2 | 8 - 9 | 896 | 64 | |
| GenBank mRNA | AK134274.1 | - | 8 - 9 | 897 | 64 | |
| AK138442.1 | - | 8 - 9 | 920 | 64 | ||
| AK146469.1 | - | 8 - 9 | 923 | 64 | ||
| AK205436.1 | - | 1 - 2 | 125 | 64 | ||
| BC011321.1 | - | 7 - 8 | 773 | 4007058 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ddx27
Genomic Map