Assay Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033196.2 | NP_001028368.2 | 12 - 13 | 3754 | 61 | |
| NM_001291162.1 | NP_001278091.1 | 5 - 6 | 689 | 61 | ||
| GenBank mRNA | AK147564.1 | - | 12 - 13 | 3757 | 61 | |
| AK147652.1 | - | 12 - 13 | 3756 | 61 | ||
| AK157301.1 | - | 5 - 6 | 692 | 61 | ||
| BC059933.1 | - | 5 - 6 | 808 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Znfx1
Genomic Map