Assay Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033196.2 | NP_001028368.2 | 13 - 14 | 3848 | 152 | |
| NM_001291162.1 | NP_001278091.1 | 6 - 7 | 783 | 152 | ||
| GenBank mRNA | AK147564.1 | - | 13 - 14 | 3851 | 152 | |
| AK147652.1 | - | 13 - 14 | 3850 | 152 | ||
| AK157301.1 | - | 6 - 7 | 786 | 152 | ||
| BC037702.1 | - | 1 - 2 | 116 | 152 | ||
| BC059933.1 | - | 6 - 7 | 902 | 152 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076757 | BC025488.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076757 | BC037702.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076757 | BC059933.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076757 | AK157301.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2076757 | NM_001033196.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 912848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 935084 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 936963 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 107153 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 28040 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 930232 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 138661 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 174441 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 93015 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 46197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 416814 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476828 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 476270 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 552883 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477515 | AF142405.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477515 | AK030277.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477515 | AK030297.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 477515 | NM_053085.2 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 22186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 40179 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 131507 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P145072 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P169759 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076757 | BC025488.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076757 | BC037702.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076757 | BC059933.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076757 | AK157301.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2076757 | NM_001033196.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144438 | BC025488.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144438 | BC037702.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144438 | BC059933.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144438 | AK157301.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P144438 | NM_001033196.2 | mouse_transcriptome |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833287 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833308 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833192 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833333 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833265 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833250 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833202 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833344 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833197 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833241 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833232 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833289 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833239 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833223 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833182 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833271 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833170 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833258 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833227 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833297 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833269 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833255 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833196 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833322 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833237 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833225 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833252 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833263 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833424 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833275 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833234 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833304 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833207 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833176 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833218 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833349 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833247 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833270 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833189 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833298 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833303 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833221 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833427 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833425 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833267 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833261 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833326 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833430 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833183 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833209 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833260 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833300 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833230 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833428 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833273 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21833243 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833435 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833439 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124943 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833431 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833444 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01187262 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124940 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833440 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01187254 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124934 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P01187257 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P21833443 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02124942 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10489863 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489854 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489859 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489853 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489855 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489860 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489865 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489857 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489862 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489861 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10489856 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1427091_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10489850 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10478847 | - | - |
| Mouse Genome 430 2.0 Array | 1427091_at | - | - |
| Mouse Genome 430 2.0 Array | 1427091_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98999_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 98999_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0200044333.mm.1 | NM_001033196.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200044333.mm.1 | NM_001033196.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0200044333.mm.1 | TC0200005181.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1427091_at | - | - |
| Mouse Expression Set 430 | 1427091_PM_at | - | - |
| Murine Genome U74v2 Set | 102965_at | - | - |
| Murine Genome U74v2 Set | 102965_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427091_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1427091_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427091_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1427091_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1219468 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2607648 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2607648 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1219468 | - | - |

Znfx1
Genomic Map