Assay Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033196.2 | NP_001028368.2 | 3 - 4 | 2402 | 86 | |
| GenBank mRNA | AK142923.1 | - | 3 - 4 | 2027 | 86 | |
| AK147564.1 | - | 3 - 4 | 2403 | 87 | ||
| AK149249.1 | - | 3 - 4 | 2021 | 86 | ||
| AK150223.1 | - | 3 - 4 | 1991 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Znfx1
Genomic Map