Assay Details
| Gene Symbol: Znfx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033196.2 | NP_001028368.2 | 6 - 7 | 2839 | 63 | |
| GenBank mRNA | AK147564.1 | - | 6 - 7 | 2841 | 63 | |
| AK147652.1 | - | 6 - 7 | 2841 | 63 | ||
| AK149249.1 | - | 6 - 7 | 2458 | 63 | ||
| AK150223.1 | - | 6 - 7 | 2428 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Znfx1
Genomic Map