Assay Details
| Gene Symbol: Slc9a8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB089793.1 | - | 9 - 10 | 874 | 70 | |
| AF482993.1 | - | 8 - 9 | 838 | 70 | ||
| AK049410.1 | - | 9 - 10 | 870 | 70 | ||
| AK143281.1 | - | 9 - 10 | 843 | 70 | ||
| AK160826.1 | - | 7 - 8 | 712 | 70 | ||
| AK173069.1 | - | 9 - 10 | 1210 | 70 | ||
| BC030879.1 | - | 9 - 10 | 873 | 4241988 | 70 | |
| BC034508.1 | - | 2 - 3 | 274 | 70 | ||
| BC058947.1 | - | 9 - 10 | 844 | 5702781 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc9a8
Genomic Map